Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002588.3 | NP_002579.2 | 1 | 2383 | |
NM_032402.1 | NP_115778.1 | 1 | 2383 | ||
GenBank mRNA | AF152337.1 | - | 1 | 2268 | |
AF152524.1 | - | 1 | 2268 | ||
BC019299.1 | - | 1 | 2382 | 2905509 | |
BC026218.1 | - | 1 | 2372 | 4153558 | |
L11371.1 | - | 1 | 75 | ||
L11373.1 | - | 1 | 2379 |