Gene Transcripts

Gene Symbol
PLCXD1
Entrez Gene ID
55344
Gene Name
phosphatidylinositol specific phospholipase C X domain containing 1
Gene Aliases
LL0XNC01-136G2.1
Chromosome Location
Chr. Y: 276324 - 303356 on Build GRCh38
UniGene ID
Hs.522568
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018390.3 NP_060860.1 7 1193
NR_028057.1 8 1176
XM_006724865.2 XP_006724928.1 8 1134
XM_006724866.2 XP_006724929.1 7 945
XM_006724867.2 XP_006724930.1 7 966
XM_011545633.2 XP_011543935.1 8 1168
GenBank mRNA AK002185.1 - 8 1178
BC005028.1 - 7 1125 3953383

View Full Product Details