Gene Transcripts

Gene Symbol
WDR11
Entrez Gene ID
55717
Gene Name
WD repeat domain 11
Gene Aliases
BRWD2, DR11, HH14, SRI1, WDR15
Chromosome Location
Chr. 10: 120851175 - 120909526 on Build GRCh38
UniGene ID
Hs.144447
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018117.11 NP_060587.8 25 3315
XM_005269962.2 XP_005270019.1 25 3032
XM_005269963.2 XP_005270020.1 25 3164
XM_005269964.2 XP_005270021.1 18 2070
XM_017016397.1 XP_016871886.1 24 3003
XM_017016398.1 XP_016871887.1 24 2933
XM_017016399.1 XP_016871888.1 24 2877
GenBank mRNA AB037772.2 - 25 3128
AF320223.1 - 25 3102
AK001368.1 - 20 2218
AK025050.1 - 8 826
AK124522.1 - 12 3498
AK299524.1 - 18 1935
AK315145.1 - 25 3093
AL137699.1 - 23 2788
BC040469.1 - 25 3132 4730308
BC071564.1 - 25 3098 30346203

View Full Product Details