Gene Transcripts
- Gene Symbol
- PSMB10
- Entrez Gene ID
- 5699
- Gene Name
- proteasome subunit beta 10
- Gene Aliases
- LMP10, MECL1, beta2i
- Chromosome Location
- Chr. 16: 67934504 - 67936877 on Build GRCh38
- UniGene ID
- Hs.9661
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002801.3 |
NP_002792.1
|
1, 2 |
183 |
|
GenBank mRNA |
AK225658.1 |
- |
1, 2 |
156 |
|
AK312208.1 |
- |
1, 2 |
143 |
|
BC017198.1 |
- |
1, 2 |
134 |
3143189 |
BC052369.1 |
- |
1, 2 |
121 |
4054165 |
BM845639.1 |
- |
1, 2 |
156 |
|
BP289065.1 |
- |
1, 2 |
183 |
|
BT019723.1 |
- |
1, 2 |
55 |
|
BT019724.1 |
- |
1, 2 |
55 |
|
Y13640.1 |
- |
1, 2 |
143 |
|
View Full Product Details