Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021035.2 | NP_066363.1 | 14 | 5909 | |
XM_006723836.1 | XP_006723899.1 | 14 | 5739 | ||
XM_011528924.1 | XP_011527226.1 | 13 | 5784 | ||
XM_017027971.1 | XP_016883460.1 | 12 | 4835 | ||
GenBank mRNA | AB037825.2 | - | 14 | 5727 | |
AK000573.1 | - | 1 | 578 | ||
AK023836.1 | - | 10 | 3661 | ||
BC062347.1 | - | 1 | 1248 |