Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022371.3 | NP_071766.2 | 6 | 1211 | |
GenBank mRNA | AJ299403.1 | - | 6 | 1100 | |
AK025998.1 | - | 6 | 1090 | ||
AK299966.1 | - | 5 | 808 | ||
AK316530.1 | - | 5 | 820 | ||
BC001085.1 | - | 6 | 1109 | 2959819 | |
BC007571.1 | - | 6 | 1100 | 3029304 | |
BC011746.1 | - | 6 | 1096 | 3343691 | |
BC018292.1 | - | 6 | 1101 | 3457259 | |
BC088368.1 | - | 5 | 838 | ||
CR457354.1 | - | 6 | 1059 |