Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001099691.2 | NP_001093161.1 | 4 | 538 | |
NM_001308158.1 | NP_001295087.1 | 4 | 708 | ||
NM_001308159.1 | NP_001295088.1 | 4 | 705 | ||
NM_003236.3 | NP_003227.1 | 4 | 541 | ||
GenBank mRNA | AF149096.1 | - | 4 | 298 | |
AF149097.1 | - | 4 | 295 | ||
AF149098.1 | - | 4 | 295 | ||
AK290151.1 | - | 4 | 352 | ||
AK309066.1 | - | 4 | 708 | ||
AK309220.1 | - | 4 | 557 | ||
AK312899.1 | - | 4 | 349 | ||
BC005308.1 | - | 4 | 349 | 3828239 | |
BT006833.1 | - | 4 | 290 | ||
K03222.1 | - | 4 | 327 | ||
M31172.1 | - | 4 | 327 | ||
X70340.1 | - | 4 | 324 |