Gene Transcripts

Gene Symbol
FBXO5
Entrez Gene ID
26271
Gene Name
F-box protein 5
Gene Aliases
EMI1, FBX5, Fbxo31
Chromosome Location
Chr. 6: 152970523 - 152983605 on Build GRCh38
UniGene ID
Hs.520506
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142522.1 NP_001135994.1 1 139
GenBank mRNA BI838832.1 - 1 170

View Full Product Details