Gene Transcripts

Gene Symbol
SECISBP2
Entrez Gene ID
79048
Gene Name
SECIS binding protein 2
Gene Aliases
SBP2
Chromosome Location
Chr. 9: 89318473 - 89367117 on Build GRCh38
UniGene ID
Hs.59804
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282688.1 NP_001269617.1 2 254
NM_001282689.1 NP_001269618.1 2 254
NM_001282690.1 NP_001269619.1 2 331
NM_024077.4 NP_076982.3 2 254
XM_005252196.2 XP_005252253.1 2 221
XM_006717282.2 XP_006717345.1 2 308
XM_011519000.2 XP_011517302.1 2 308
XM_011519001.1 XP_011517303.1 2 308
XM_011519002.1 XP_011517304.1 2 308
XM_011519003.1 XP_011517305.1 2 308
XM_017015122.1 XP_016870611.1 2 221
XM_017015123.1 XP_016870612.1 2 221
XM_017015124.1 XP_016870613.1 2 221
GenBank mRNA AB208940.1 - 2 212
AF380995.1 - 2 207
AK090608.1 - 2 221
AK290182.1 - 2 221
AK299161.1 - 2 342
AK302852.1 - 2 252
AL136881.1 - 2 208
BC001189.1 - 2 208
BC036109.1 - 2 216 5296292
BC068466.1 - 2 189
BC099900.1 - 2 190
BP219148.1 - 2 254
DR002073.1 - 2 247

View Full Product Details