Gene Transcripts
- Gene Symbol
- PAAF1
- Entrez Gene ID
- 80227
- Gene Name
- proteasomal ATPase associated factor 1
- Gene Aliases
- PAAF, Rpn14, WDR71
- Chromosome Location
- Chr. 11: 73876699 - 73927736 on Build GRCh38
- UniGene ID
- Hs.525017
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001267803.1 |
NP_001254732.1
|
9, 10 |
932 |
|
NM_001267804.1 |
NP_001254733.1
|
9, 10 |
923 |
|
NM_001267805.1 |
NP_001254734.1
|
9, 10 |
919 |
|
NM_001267806.1 |
NP_001254735.1
|
8, 9 |
806 |
|
NM_025155.2 |
NP_079431.1
|
8, 9 |
863 |
|
XM_005274308.2 |
XP_005274365.2
|
6, 7 |
1455 |
|
XM_005274309.1 |
XP_005274366.1
|
7, 8 |
722 |
|
XM_017018357.1 |
XP_016873846.1
|
9, 10 |
851 |
|
XM_017018358.1 |
XP_016873847.1
|
9, 10 |
856 |
|
XM_017018359.1 |
XP_016873848.1
|
6, 7 |
1414 |
|
GenBank mRNA |
AF087895.1 |
- |
9, 10 |
857 |
|
AK021910.1 |
- |
8, 9 |
844 |
|
AK225520.1 |
- |
8, 9 |
831 |
|
AK298258.1 |
- |
8, 9 |
806 |
|
AK309133.1 |
- |
6, 7 |
1070 |
|
AK316394.1 |
- |
8, 9 |
1077 |
|
BC006142.2 |
- |
8, 9 |
816 |
3937356 |
BC021541.1 |
- |
8, 9 |
827 |
3942680 |
BC028628.1 |
- |
9, 10 |
897 |
5263875 |
View Full Product Details