Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025163.3 | NP_079439.2 | 3 | 1043 | |
XM_011513190.1 | XP_011511492.1 | 4 | 1634 | ||
XM_011513192.2 | XP_011511494.1 | 4 | 1160 | ||
XM_017007240.1 | XP_016862729.1 | 3 | 2001 | ||
XM_017007241.1 | XP_016862730.1 | 4 | 3378 | ||
XM_017007242.1 | XP_016862731.1 | 3 | 3070 | ||
XM_017007243.1 | XP_016862732.1 | 3 | 962 | ||
GenBank mRNA | BC018804.1 | - | 3 | 924 | |
BC044640.1 | - | 3 | 925 | 4871581 |