Gene Transcripts

Gene Symbol
MFSD7
Entrez Gene ID
84179
Gene Name
major facilitator superfamily domain containing 7
Gene Aliases
LP2561
Chromosome Location
Chr. 4: 676205 - 689468 on Build GRCh38
UniGene ID
Hs.567612
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001294341.1 NP_001281270.1 1, 2 450
NM_032219.3 NP_115595.2 1, 2 450
XM_005272297.1 XP_005272354.1 1, 2 477
XM_005272299.4 XP_005272356.1 1, 2 477
XM_006713919.1 XP_006713982.1 1, 2 477
XM_011513573.2 XP_011511875.1 1, 2 477
XM_011513574.1 XP_011511876.1 1, 2 477
XM_017008700.1 XP_016864189.1 1, 2 477
GenBank mRNA AK292010.1 - 1, 2 348
AY358401.1 - 1, 2 152
BC030246.1 - 1, 2 193 5216645

View Full Product Details