Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004292.2 | NP_004283.2 | 7 | 1425 | |
XM_011545399.2 | XP_011543701.1 | 7 | 1774 | ||
XM_011545401.2 | XP_011543703.1 | 7 | 1299 | ||
GenBank mRNA | AB209445.1 | - | 4 | 1328 | |
AK297757.1 | - | 6 | 1121 | ||
BC014417.1 | - | 7 | 1315 | 4549333 | |
KU178481.1 | - | 7 | 1298 | ||
L36463.1 | - | 7 | 1425 |