Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001126060.1 | NP_001119532.2 | 2 | 2989 | |
NM_001164757.1 | NP_001158229.1 | 10 | 1879 | ||
NM_014697.2 | NP_055512.1 | 10 | 1894 | ||
GenBank mRNA | AB007933.2 | - | 10 | 1894 | |
AF037070.1 | - | 2 | 448 | ||
AY841899.1 | - | 2 | 2989 | ||
BC041382.2 | - | 10 | 1896 | ||
BC112295.1 | - | 10 | 1595 | 8327763 | |
BC143771.1 | - | 10 | 1580 | 9052285 | |
BC143780.1 | - | 11 | 1693 |