Gene Transcripts

Gene Symbol
Tbc1d10c
Entrez Gene ID
108995
Gene Name
TBC1 domain family, member 10c
Gene Aliases
1810062O14Rik, AI428527
Chromosome Location
Chr. 19: 4184357 - 4191195 on Build GRCm38
UniGene ID
Mm.288312
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178650.3 NP_848765.2 2 164
GenBank mRNA AK040487.1 - 2 165
AK075803.1 - 2 151
AK088119.1 - 229
AK131177.1 - 1 145
AK146426.1 - 2 158
AK153836.1 - 2 162
AK154444.1 - 1 142
BC094901.1 - 1 139
BQ829173.1 - 2 400

View Full Product Details