Gene Transcripts

Gene Symbol
Vmn1r44
Entrez Gene ID
113854
Gene Name
vomeronasal 1 receptor 44
Gene Aliases
V1RA10, V1rb11, V1rb4
Chromosome Location
Chr. 6: 89892161 - 89894206 on Build GRCm38
UniGene ID
Mm.377151
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053227.2 NP_444457.2 1 809
GenBank mRNA BC107184.1 - 1 809
BC107185.1 - 1 809
BC151088.1 - 1 906 9088001

View Full Product Details