Gene Transcripts
- Gene Symbol
- Mest
- Entrez Gene ID
- 17294
- Gene Name
- mesoderm specific transcript
- Gene Aliases
- AA408879, AI256745, Peg1
- Chromosome Location
- Chr. 6: 30733506 - 30748466 on Build GRCm38
- UniGene ID
- Mm.335639
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001252292.1 |
NP_001239221.1
|
6, 7 |
834 |
|
NM_001252293.1 |
NP_001239222.1
|
5, 6 |
555 |
|
NM_008590.2 |
NP_032616.1
|
5, 6 |
733 |
|
GenBank mRNA |
AF482999.1 |
- |
6, 7 |
555 |
|
AK031718.1 |
- |
5, 6 |
732 |
|
AK032881.1 |
- |
5, 6 |
732 |
|
AK034949.1 |
- |
5, 6 |
731 |
|
AK146457.1 |
- |
6, 7 |
834 |
|
AK168714.1 |
- |
5, 6 |
731 |
|
AK168732.1 |
- |
5, 6 |
731 |
|
AK168743.1 |
- |
5, 6 |
731 |
|
AK168995.1 |
- |
5, 6 |
731 |
|
AK169095.1 |
- |
5, 6 |
731 |
|
AK169266.1 |
- |
5, 6 |
731 |
|
BC006639.1 |
- |
5, 6 |
716 |
3482928 |
CF916557.1 |
- |
5, 6 |
547 |
|
D16262.1 |
- |
5, 6 |
720 |
|
View Full Product Details