Gene Transcripts

Gene Symbol
Nr4a3
Entrez Gene ID
18124
Gene Name
nuclear receptor subfamily 4, group A, member 3
Gene Aliases
AI573420, CHN, CSMF, MINOR, NOR-1, Nor1, TEC
Chromosome Location
Chr. 4: 48044637 - 48086447 on Build GRCm38
UniGene ID
Mm.247261
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB221628.1 - 3 1244
AF191211.1 - 3 1244
BC068150.1 - 5 1831
BC128307.1 - 3 1239
BC128308.1 - 3 1240

View Full Product Details