Gene Transcripts

Gene Symbol
Prkcq
Entrez Gene ID
18761
Gene Name
protein kinase C, theta
Gene Aliases
A130035A12Rik, AW494342, PKC-0, PKC-theta, PKCtheta, Pkcq
Chromosome Location
Chr. 2: 11172382 - 11301226 on Build GRCm38
UniGene ID
Mm.329993
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008859.2 NP_032885.1 3 267
GenBank mRNA AK037664.1 - 3 339
AK085546.1 - 3 537
BC138552.1 - 2 198 8861572
BC145769.1 - 2 198 40131072
D11091.1 - 3 267

View Full Product Details