Gene Transcripts

Gene Symbol
Pou2f1
Entrez Gene ID
18986
Gene Name
POU domain, class 2, transcription factor 1
Gene Aliases
2810482H01Rik, NF-A1, Oct-1, Oct1, Otf-1, Otf1
Chromosome Location
Chr. 1: 165865150 - 166002672 on Build GRCm38
UniGene ID
Mm.245261
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011137.3 NP_035267.2 12 1329
NM_198932.3 NP_945150.2 12 1329
NM_198933.3 NP_945151.2 12 1401
NM_198934.3 NP_945152.2 12 1401
GenBank mRNA AF508939.1 - 12 1310
AJ296212.1 - 12 1353
AJ489474.1 - 12 1401
AJ489475.1 - 11 1218
AK028237.1 - 12 1331
AY177625.1 - 12 1310
BC066201.1 - 12 1306
S65461.1 - 11 1229
X51958.1 - 3 336
X56230.1 - 11 1406
X68362.1 - 12 1302
X68363.1 - 12 1300
X68364.1 - 12 1316
X70324.1 - 10 1161
X70325.1 - 2 142

View Full Product Details