Gene Transcripts

Gene Symbol
Rab18
Entrez Gene ID
19330
Gene Name
RAB18, member RAS oncogene family
Gene Aliases
AA959686
Chromosome Location
Chr. 18: 6765014 - 6791606 on Build GRCm38
UniGene ID
Mm.132802
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278447.1 NP_001265376.1 7 723
NM_181070.6 NP_851415.1 7 705
GenBank mRNA AB232612.1 - 7 607
AK028155.1 - 3 306
AK045514.1 - 7 668
AK140901.1 - 7 673
AK143673.1 - 7 681
AK146177.1 - 7 630
AK146397.1 - 7 630
AK154920.1 - 7 682
BC056351.1 - 7 659 6807284
L04966.1 - 7 611

View Full Product Details