Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252313.1 | NP_001239242.1 | 41 | 6678 | |
NM_011308.3 | NP_035438.3 | 41 | 6477 | ||
GenBank mRNA | AB093281.1 | - | 23 | 4418 | |
AK034988.1 | - | 1 | 2175 | ||
AK038502.1 | - | 1 | 125 | ||
AK052753.1 | - | 1 | 498 | ||
AK137632.1 | - | 15 | 2796 | ||
BC086657.1 | - | 41 | 6443 | 30545994 | |
L78294.1 | - | 20 | 3482 | ||
U22016.1 | - | 7 | 1232 | ||
U35312.1 | - | 40 | 6721 |