Gene Transcripts
- Gene Symbol
- Sel1l
- Entrez Gene ID
- 20338
- Gene Name
- sel-1 suppressor of lin-12-like (C. elegans)
- Gene Aliases
- AW493766, Sel1h, mKIAA4137
- Chromosome Location
- Chr. 12: 91806043 - 91849157 on Build GRCm38
- UniGene ID
- Mm.250605
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039089.1 |
NP_001034178.1
|
20, 21 |
2289 |
|
NM_011344.2 |
NP_035474.1
|
20, 21 |
2139 |
|
GenBank mRNA |
AF063095.1 |
- |
20, 21 |
2088 |
|
AK005023.2 |
- |
20, 21 |
2262 |
|
AK146022.1 |
- |
19, 20 |
2877 |
|
AK160936.1 |
- |
20, 21 |
2097 |
|
AK220502.1 |
- |
20, 21 |
2289 |
|
BC026816.1 |
- |
20, 21 |
2111 |
4457637 |
BC049959.1 |
- |
15, 16 |
1501 |
|
BC057452.1 |
- |
6, 7 |
724 |
|
View Full Product Details