Gene Transcripts
- Gene Symbol
- St6galnac4
- Entrez Gene ID
- 20448
- Gene Name
- ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 4
- Gene Aliases
- SIAT7-D, ST6GalNAcIV, Siat7d
- Chromosome Location
- Chr. 2: 32587078 - 32599696 on Build GRCm38
- UniGene ID
- Mm.27446
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001276425.1 |
NP_001263354.1
|
3, 4 |
442 |
|
NM_011373.3 |
NP_035503.1
|
2, 3 |
795 |
|
GenBank mRNA |
AJ007310.1 |
- |
3, 4 |
437 |
|
AK007601.1 |
- |
3, 4 |
425 |
|
AK085730.1 |
- |
3, 4 |
433 |
|
AK144884.1 |
- |
3, 4 |
433 |
|
AK153036.1 |
- |
3, 4 |
402 |
|
AK155956.1 |
- |
3, 4 |
433 |
|
AK159712.1 |
- |
2, 3 |
786 |
|
AK161559.1 |
- |
3, 4 |
427 |
|
BC044734.2 |
- |
2, 3 |
741 |
|
BC056451.1 |
- |
2, 3 |
248 |
5685866 |
BC086784.1 |
- |
|
383 |
6844232 |
Y15779.1 |
- |
3, 4 |
419 |
|
Y15780.1 |
- |
2, 3 |
431 |
|
View Full Product Details