Gene Transcripts

Gene Symbol
Spp1
Entrez Gene ID
20750
Gene Name
secreted phosphoprotein 1
Gene Aliases
2AR, Apl-1, BNSP, BSPI, Bsp, ETA-1, Eta, OP, Opn, Opnl, Ric, Spp-1
Chromosome Location
Chr. 5: 104435111 - 104441053 on Build GRCm38
UniGene ID
Mm.288474
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204201.1 NP_001191130.1 5, 6 315
NM_001204202.1 NP_001191131.1 4, 5 312
NM_001204203.1 NP_001191132.1 5, 6 263
NM_001204233.1 NP_001191162.1 3, 4 357
NM_009263.3 NP_033289.2 4, 5 260
GenBank mRNA AF515708.1 - 3, 4 160
AK133555.1 - 4, 5 251
AK145099.1 - 4, 5 254
AK149588.1 - 4, 5 251
AK150819.1 - 4, 5 251
AK150845.1 - 4, 5 251
AK151069.1 - 4, 5 221
AK151562.1 - 4, 5 251
AK153089.1 - 4, 5 251
AK154908.1 - 4, 5 251
AK160540.1 - 4, 5 254
AK165393.1 - 4, 5 251
AK165965.1 - 4, 5 303
AK166395.1 - 4, 5 254
AK171158.1 - 4, 5 251
BC002113.1 - 4, 5 257 3489075
BC020355.1 - 4, 5 240
BC029991.1 - 4, 5 240
BC057858.1 - 4, 5 233 5294167
BC080720.1 - 4, 5 233 5252869
CT010357.1 - 3, 4 160
J04806.1 - 4, 5 229
S78177.1 - 2, 3 115
X13986.1 - 3, 4 175
X16151.1 - 3, 4 382

View Full Product Details