Gene Transcripts

Gene Symbol
Rpp40
Entrez Gene ID
208366
Gene Name
ribonuclease P 40 subunit
Gene Aliases
D8Bwg1265e, Rnasep1
Chromosome Location
Chr. 13: 35895104 - 35911565 on Build GRCm38
UniGene ID
Mm.35545
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145938.4 NP_666050.3 3, 4 353
GenBank mRNA AK048810.1 - 353
BC024607.1 - 3, 4 298 4236826

View Full Product Details