Gene Transcripts

Gene Symbol
Trim30d
Entrez Gene ID
209387
Gene Name
tripartite motif-containing 30D
Gene Aliases
AI451617, TRIM30-3, Trim79
Chromosome Location
Chr. 7: 104470014 - 104507849 on Build GRCm38
UniGene ID
Mm.277377
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167828.1 NP_001161300.1 5 593
NM_199146.2 NP_954597.2 6 1069
GenBank mRNA AK136456.1 - 5 597
AK164938.1 - 5 592
BC056631.1 - 6 1045 5035734

View Full Product Details