Gene Transcripts

Gene Symbol
Tbl1x
Entrez Gene ID
21372
Gene Name
transducin (beta)-like 1 X-linked
Gene Aliases
5330429M20, Tbl1
Chromosome Location
Chr. X: 77510547 - 77662985 on Build GRCm38
UniGene ID
Mm.258476
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020601.2 NP_065626.1 5 429
GenBank mRNA AK030547.1 - 5 644
AK036996.1 - 6 508
AK038674.1 - 5 430
BC043105.1 - 5 552 6406579

View Full Product Details