Gene Transcripts

Gene Symbol
Pomt2
Entrez Gene ID
217734
Gene Name
protein-O-mannosyltransferase 2
Gene Aliases
A830009D15Rik, AW046274
Chromosome Location
Chr. 12: 87106861 - 87147902 on Build GRCm38
UniGene ID
Mm.388249
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153415.3 NP_700464.2 9, 10 1601
GenBank mRNA AF246235.1 - 9, 10 1205
AK043573.1 - 9, 10 1302
AK043713.1 - 9, 10 1143
AK050915.1 - 9, 10 1170
AK076813.1 - 9, 10 1603
AY090481.1 - 9, 10 1601
AY090482.1 - 9, 10 1201
BC052045.1 - 9, 10 1173 6402328
BC062965.1 - 9, 10 1173
BC079589.1 - 9, 10 1171

View Full Product Details