Gene Transcripts

Gene Symbol
Vdac3
Entrez Gene ID
22335
Gene Name
voltage-dependent anion channel 3
Gene Aliases
Chromosome Location
Chr. 8: 22577075 - 22593813 on Build GRCm38
UniGene ID
Mm.227704
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198998.1 NP_001185927.1 7 517
NM_011696.2 NP_035826.1 6 514
GenBank mRNA AK002296.1 - 6 462
AK083785.1 - 7 445
AK146957.1 - 7 470
AK150741.1 - 6 456
AK150828.1 - 7 445
AK159431.1 - 6 456
AK161283.1 - 6 443
AK167781.1 - 6 459
AK168815.1 - 7 462
AK169099.1 - 7 470
AK169228.1 - 6 442
BC004743.1 - 6 412 3583800
BC089336.1 - 7 434 30357677
U30839.1 - 6 459

View Full Product Details