Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_181750.2 | NP_861415.2 | 10 | 1174 | |
GenBank mRNA | AK141150.1 | - | 8 | 1028 | |
AK142916.1 | - | 2 | 169 | ||
AK144098.1 | - | 8 | 1048 | ||
BC032160.1 | - | 1 | 66 | ||
BC049181.1 | - | 10 | 1156 | ||
BC137772.1 | - | 9 | 919 | 8860791 | |
BC144899.1 | - | 9 | 919 |