Gene Transcripts

Gene Symbol
Ints3
Entrez Gene ID
229543
Gene Name
integrator complex subunit 3
Gene Aliases
C77668
Chromosome Location
Chr. 3: 90391380 - 90433636 on Build GRCm38
UniGene ID
Mm.268564
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145540.3 NP_663515.2 4 1025
NM_178876.3 NP_849207.2 4 1025
GenBank mRNA AK033296.1 - 4 692
AK040147.1 - 4 1015
AK146699.1 - 4 919
AK147865.1 - 4 1004
AK188601.1 - 1 18
BC055344.1 - 4 560 6827687

View Full Product Details