Gene Transcripts

Gene Symbol
Dnajc11
Entrez Gene ID
230935
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 11
Gene Aliases
E030019A03Rik
Chromosome Location
Chr. 4: 151933720 - 151981959 on Build GRCm38
UniGene ID
Mm.21353
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172704.3 NP_766292.2 8 866
GenBank mRNA AK053156.1 - 8 848
AK089983.1 - 8 858
AK156793.1 - 6 2958
AK171781.1 - 8 847
AK172542.1 - 8 847
AK181713.1 - 1 83
AK186483.1 - 1 83
BC075619.1 - 7 719
BC079852.1 - 8 836
BC085257.1 - 8 840 5414963

View Full Product Details