Gene Transcripts

Gene Symbol
Slfn10-ps
Entrez Gene ID
237887
Gene Name
schlafen 10, pseudogene
Gene Aliases
Slfn10, mFLJ00257
Chromosome Location
Chr. 11: 83028123 - 83040533 on Build GRCm38
UniGene ID
Mm.341645
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK041394.1 - 4 1782
AK144698.1 - 5 2375
AK220241.1 - 5 2499
AY261802.1 - 5 2374
AY261803.1 - 5 2343
BC115828.1 - 1266

View Full Product Details