Gene Transcripts

Gene Symbol
Slc30a8
Entrez Gene ID
239436
Gene Name
solute carrier family 30 (zinc transporter), member 8
Gene Aliases
C820002P14Rik, ZnT-8, ZnT8
Chromosome Location
Chr. 15: 52295553 - 52335733 on Build GRCm38
UniGene ID
Mm.208831
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172816.3 NP_766404.1 3 640
GenBank mRNA AK050482.1 - 3 541
AK050494.1 - 3 640
BC117544.1 - 3 472 40105617
BC118521.1 - 3 469 40105622

View Full Product Details