Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172931.3 | NP_766519.1 | 2, 3 | 231 | |
GenBank mRNA | AB510446.1 | - | 3, 4 | 333 | |
AB510447.1 | - | 833 | |||
AK046229.1 | - | 261 | |||
BC117805.2 | - | 2, 3 | 231 | 40092308 |