Gene Transcripts
- Gene Symbol
- B3gat2
- Entrez Gene ID
- 280645
- Gene Name
- beta-1,3-glucuronyltransferase 2 (glucuronosyltransferase S)
- Gene Aliases
- GlcAT-S, Glcats
- Chromosome Location
- Chr. 1: 23761926 - 23847865 on Build GRCm38
- UniGene ID
- Mm.471661
- Species
- Mus musculus
- Gene Symbol
- Smap1
- Entrez Gene ID
- 98366
- Gene Name
- small ArfGAP 1
- Gene Aliases
- 1700056O10Rik, 4921514B13Rik, 4921525H11Rik, AI462175
- Chromosome Location
- Chr. 1: 23844825 - 23922450 on Build GRCm38
- UniGene ID
- Mm.329963
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK132868.1 |
- |
11 |
1797 |
|
BE631392.1 |
- |
1 |
427 |
|
View Full Product Details