Gene Transcripts

Gene Symbol
Ranbp17
Entrez Gene ID
66011
Gene Name
RAN binding protein 17
Gene Aliases
Chromosome Location
Chr. 11: 33211794 - 33514750 on Build GRCm38
UniGene ID
Mm.383775
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023146.2 NP_075635.1 21 2424
GenBank mRNA AJ271458.1 - 21 2322
AK016583.1 - 21 2324
AK050150.1 - 17 1875
BC033503.1 - 22 2434

View Full Product Details