Gene Transcripts

Gene Symbol
Peli1
Entrez Gene ID
67245
Gene Name
pellino 1
Gene Aliases
2810468L03Rik, A930031K15Rik, AA409794, AI586297, D11Ertd676e
Chromosome Location
Chr. 11: 21091324 - 21150327 on Build GRCm38
UniGene ID
Mm.28957
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023324.2 NP_075813.2 7 1316
GenBank mRNA AF302503.1 - 6 1538
AK009945.1 - 1 125
AK013388.1 - 1 1086
AK076458.1 - 1 38
AK151660.1 - 7 1317
AK152900.1 - 7 1317
AK152997.1 - 7 1316
AK153372.1 - 7 1317
BC016515.1 - 6 915 3600130

View Full Product Details