Gene Transcripts

Gene Symbol
Mansc1
Entrez Gene ID
67729
Gene Name
MANSC domain containing 1
Gene Aliases
9130403P13Rik
Chromosome Location
Chr. 6: 134609207 - 134632488 on Build GRCm38
UniGene ID
Mm.57648
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026345.4 NP_080621.1 4 1201
GenBank mRNA AK002644.1 - 4 1110
AK018635.1 - 4 1092
AK018660.1 - 4 1114
AK033526.1 - 4 1113
AK033557.1 - 4 1202
AK046837.1 - 4 1090

View Full Product Details