Gene Transcripts

Gene Symbol
Ccdc80
Entrez Gene ID
67896
Gene Name
coiled-coil domain containing 80
Gene Aliases
2610001E17Rik, DRO1, Ssg1, Urb
Chromosome Location
Chr. 16: 45093407 - 45127924 on Build GRCm38
UniGene ID
Mm.181074
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026439.2 NP_080715.2 2 675
GenBank mRNA AB075019.1 - 2 564
AK032359.1 - 2 646
AK047367.1 - 1 1736
AK132178.1 - 2 675

View Full Product Details