Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026439.2 | NP_080715.2 | 2 | 458 | |
GenBank mRNA | AB075019.1 | - | 2 | 347 | |
AK032359.1 | - | 2 | 429 | ||
AK047367.1 | - | 1 | 1953 | ||
AK132178.1 | - | 2 | 458 | ||
AK144183.1 | - | 3 | 682 | ||
BC058751.1 | - | 2 | 217 | ||
BC096487.1 | - | 3 | 648 |