Gene Transcripts

Gene Symbol
Klhl30
Entrez Gene ID
70788
Gene Name
kelch-like 30
Gene Aliases
4631423F02Rik
Chromosome Location
Chr. 1: 91351073 - 91362404 on Build GRCm38
UniGene ID
Mm.68617
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027551.2 NP_081827.1 7 1551
GenBank mRNA AK014532.1 - 3 418
AK086025.1 - 7 1552
AK146580.1 - 7 1608
AK148398.1 - 7 1560
BC115428.1 - 6 1494 40094085
BC115429.1 - 6 1494 40094088

View Full Product Details