Gene Transcripts

Gene Symbol
Tchhl1
Entrez Gene ID
71325
Gene Name
trichohyalin-like 1
Gene Aliases
5430400H23Rik, AA589420, S100a17, Thhl1
Chromosome Location
Chr. 3: 93468734 - 93471980 on Build GRCm38
UniGene ID
Mm.68064
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027762.3 NP_082038.2 3 959
GenBank mRNA AA500450.1 - 1 189
AK017244.1 - 3 960

View Full Product Details