Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001032413.1 | NP_001027585.1 | 8 | 878 | |
NM_001032414.1 | NP_001027586.1 | 7 | 808 | ||
NM_001289600.1 | NP_001276529.1 | 7 | 780 | ||
NM_001289601.1 | NP_001276530.1 | 9 | 1021 | ||
NM_028460.2 | NP_082736.1 | 9 | 1012 | ||
NM_152799.2 | NP_690012.2 | 9 | 977 | ||
GenBank mRNA | AF440279.1 | - | 9 | 1002 | |
AF444274.1 | - | 7 | 802 | ||
AF461685.1 | - | 6 | 735 | ||
AK042308.1 | - | 8 | 1036 | ||
AK053551.1 | - | 8 | 869 | ||
AK141082.1 | - | 775 | |||
AK161049.1 | - | 9 | 1018 | ||
BC042490.1 | - | 8 | 827 | 4953584 |