Gene Transcripts

Gene Symbol
Mospd2
Entrez Gene ID
76763
Gene Name
motile sperm domain containing 2
Gene Aliases
2410013I23Rik
Chromosome Location
Chr. X: 164936169 - 164980375 on Build GRCm38
UniGene ID
Mm.291758
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290523.1 NP_001277452.1 5 488
NM_001290524.1 NP_001277453.1 5 488
NM_029730.4 NP_084006.2 5 488
GenBank mRNA AK010480.1 - 5 410
AK031405.1 - 5 413
AK035985.1 - 5 448
AK039893.1 - 5 439
AK135188.1 - 5 443
AK156572.1 - 5 431
BC026425.1 - 5 414 4223175

View Full Product Details