Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023668.2 | NP_076157.2 | 6 | 810 | |
GenBank mRNA | AF290472.1 | - | 6 | 742 | |
AF323918.1 | - | 6 | 830 | ||
AK011168.1 | - | 6 | 720 | ||
AK146029.1 | - | 6 | 739 | ||
BC021434.1 | - | 706 | 5029832 | ||
BC046796.1 | - | 6 | 741 | 5718032 |