Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033588.4 | NP_291066.1 | 1 | 1240 | |
GenBank mRNA | AK043161.1 | - | 1 | 1355 | |
AK043491.1 | - | 1 | 1317 | ||
AY013799.1 | - | 1 | 1240 | ||
BC057404.1 | - | 1 | 1340 | 6405582 |