Gene Transcripts

Gene Symbol
Pabpc5
Entrez Gene ID
93728
Gene Name
poly(A) binding protein, cytoplasmic 5
Gene Aliases
C820015E17
Chromosome Location
Chr. X: 119927196 - 119930165 on Build GRCm38
UniGene ID
Mm.125979
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053114.2 NP_444344.1 2 347
GenBank mRNA AK050551.1 - 2 349
BC107362.1 - 1 248 40056131
BC107363.2 - 1 248 40056132

View Full Product Details