Gene Transcripts
- Gene Symbol
- Rell1
- Entrez Gene ID
- 100532
- Gene Name
- RELT-like 1
- Gene Aliases
- AA536743
- Chromosome Location
- Chr. 5: 63888667 - 63968900 on Build GRCm38
- UniGene ID
- Mm.243632
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145923.4 |
NP_666035.2
|
3, 4 |
562 |
|
GenBank mRNA |
AK133845.1 |
- |
4, 5 |
582 |
|
AK140108.1 |
- |
3, 4 |
562 |
|
AK159944.1 |
- |
3, 4 |
488 |
|
AK161061.1 |
- |
3, 4 |
488 |
|
AK182372.1 |
- |
2, 3 |
121 |
|
AK202598.1 |
- |
2, 3 |
121 |
|
BC024679.1 |
- |
|
11 |
|
BC031198.1 |
- |
3, 4 |
531 |
5059693 |
BC066137.1 |
- |
3, 4 |
541 |
30012041 |
BC066160.1 |
- |
3, 4 |
541 |
30012671 |
View Full Product Details